missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Antibody (ACTN2/3294) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications
Specifications
| Antigen | Alpha Actinin 2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | ACTN2/3294 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?