missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Actinin 1 Recombinant Rabbit Monoclonal Antibody (6M2L3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557907
This item is not returnable.
View return policy
Description
Immunogen sequence: KLASDLLEWI RRTIPWLENR VPENTMHAMQ QKLEDFRDYR RLHKPPKVQE KCQLEINFNT LQTKLRLSNR PAFMPSEGRM VSDINNAWGC LEQVEKGYEE.
Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| alpha Actinin 1 | |
| Recombinant Monoclonal | |
| 1.57 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P12814, Q7TPR4 | |
| ACTN1 | |
| A synthetic peptide corresponding to a sequence within amino acids 281-380 of human alpha-Actinin (NP_001093.1). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6M2L3 | |
| Unconjugated | |
| ACTN1 | |
| 3110023F10Rik; actinin; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; Actn1; Actn1a; alpha actinin 1a; alpha-actinin; alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; fibroblast alpha actinin; FLJ40884; FLJ54432; hypothetical protein; hypothetical protein LOC524770; I79_004057; non-muscle alpha-actinin 1; non-muscle alpha-actinin-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 109711, 87 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction