missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Actinin 1 Polyclonal Antibody

alpha Actinin 1 Polyclonal Antibody
Brand: Invitrogen PA5102370
This item is not returnable.
View return policy
Description
alpha Actinin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| alpha Actinin 1 | |
| Polyclonal | |
| Unconjugated | |
| ACTN1 | |
| 3110023F10Rik; actinin; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; Actn1; Actn1a; alpha actinin 1a; alpha-actinin; alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; fibroblast alpha actinin; FLJ40884; FLJ54432; hypothetical protein; hypothetical protein LOC524770; I79_004057; non-muscle alpha-actinin 1; non-muscle alpha-actinin-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109711, 81634, 87 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12814, Q7TPR4, Q9Z1P2 | |
| ACTN1 | |
| A synthesized peptide derived from human ACTN1(Accession P12814), corresponding to amino acid residues K31-E81. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction