missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Actinin 1 Antibody (1D10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59465APC
This item is not returnable.
View return policy
Description
alpha-Actinin 1 Monoclonal antibody specifically detects alpha-Actinin 1 in Human samples. It is validated for Flow Cytometry
Specifications
| alpha-Actinin 1 | |
| Monoclonal | |
| APC | |
| actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
| Recombinant human alpha-Actinin 1 (1-249aa) purified from E. coli | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers | |
| 87 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 1D10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction