missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-2C Adrenergic R/ADRA2C Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | alpha-2C Adrenergic R/ADRA2C |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18360917
|
Novus Biologicals
NBP3-15504-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18357766
|
Novus Biologicals
NBP3-15504-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-2C Adrenergic R/ADRA2C Polyclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western BlotSpecifications
| alpha-2C Adrenergic R/ADRA2C | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 240-380 of human alpha-2C Adrenergic R/ADRA2C (NP_000674.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS, 50% glycerol, pH7.3 | |
| 152 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title