missing translation for 'onlineSavingsMsg'
Learn More

alpha-2C Adrenergic R/ADRA2C Antibody (S330A-80), Novus Biologicals™

Mouse Monoclonal Antibody

£177.00

Specifications

Antigen alpha-2C Adrenergic R/ADRA2C
Clone S330A-80
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Applications Western Blot, Immunocytochemistry, Immunofluorescence
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793114
View Documents
Novus Biologicals
NBP2-22397-0.025mg
0.025 mg
£177.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha-2C Adrenergic R/ADRA2C Monoclonal specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

alpha-2C Adrenergic R/ADRA2C
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
GPCR, Neuroscience
P18825
152
Synthetic peptide amino acids 442- 462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
S330A-80
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Monoclonal
Purified
RUO
Human, Mouse
ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
ADRA2C
IgG1
Protein G purified
Detects 50kDa or larger (possibly due to dimerization). Does not cross-react with other adrenergic receptors.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.