missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-2C Adrenergic R/ADRA2C Antibody (S330A-80), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22397JF646
This item is not returnable.
View return policy
Description
alpha-2C Adrenergic R/ADRA2C Monoclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| alpha-2C Adrenergic R/ADRA2C | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| S330A-80 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR | |
| Synthetic peptide amino acids 442- 462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 152 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction