missing translation for 'onlineSavingsMsg'
Learn More

alpha-2C Adrenergic R/ADRA2C Antibody (S330A-80), DyLight 755, Novus Biologicals™

Product Code. 18424955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424955 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424955 Supplier Novus Biologicals Supplier No. NBP222397IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-2C Adrenergic R/ADRA2C Monoclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2C Adrenergic R/ADRA2C
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone S330A-80
Conjugate DyLight 755
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
Host Species Mouse
Immunogen Synthetic peptide amino acids 442- 462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 152
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.