missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha-2b Adrenergic Receptor Monoclonal Antibody (5G10)
GREENER_CHOICE

Product Code. 11538213
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538213 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538213 Supplier Invitrogen™ Supplier No. MA112721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

A suggested positive control for this product is MDCK cells transfected with rat alpha2B adrenergic receptors.

Alpha-2-adrenergic receptors are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system. Alpha 2B adrenergic receptor subtype was observed to associate with eIF-2B, a guanine nucleotide exchange protein that functions in regulation of translation. A polymorphic variant of the alpha2B subtype, which lacks 3 glutamic acids from a glutamic acid repeat element, was identified to have decreased G protein-coupled receptor kinase-mediated phosphorylation and desensitization; this polymorphic form is also associated with reduced basal metabolic rate in obese subjects and may therefore contribute to the pathogenesis of obesity. Alpha 2B adrenergic receptor gene contains no introns in either its coding or untranslated sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2b Adrenergic Receptor
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5G10
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene ADRA2B
Gene Accession No. P19328, P30545
Gene Alias [a]2B; ADA2B; Adra2b; Adra-2b; ADRA2B adrenergic, alpha-2B-, receptor; ADRA2L1; ADRA2RL1; ADRARL1; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; adrenoceptor alpha 2B; alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; alpha2B; alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; alpha-2B adrenoreceptor; ALPHA2BAR; alpha-2BAR; alpha2-C2; FAME2; G-protein coupled receptor; subtype alpha2-C2
Gene Symbols ADRA2B
Host Species Mouse
Immunogen Synthetic peptide corresponding to the third intracellular loop of rat alpha 2b adrenergic receptor.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11552, 24174
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.