missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-2B Adrenergic R/ADRA2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £256.00
Specifications
| Antigen | alpha-2B Adrenergic R/ADRA2B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680652
|
Novus Biologicals
NBP2-97424-100ul |
100 μg |
£256.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611274
|
Novus Biologicals
NBP2-97424-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-2B Adrenergic R/ADRA2B Polyclonal antibody specifically detects alpha-2B Adrenergic R/ADRA2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| alpha-2B Adrenergic R/ADRA2B | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, GPCR, Immunology, Innate Immunity | |
| PBS (pH 7.0) | |
| 151 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor | |
| Produced in rabbits immunized with E. coli-derived Human alpha-2B Adrenergic R/ADRA2B fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title