missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-7339-0.1mg
This item is not returnable.
View return policy
Description
alpha 2-Macroglobulin Polyclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| alpha 2-Macroglobulin | |
| Polyclonal | |
| HRP | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| alpha 2-Macroglobulin [Human Plasma] (Uniprot: P01023) | |
| 0.1 mg | |
| Neuroscience | |
| 2 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:1000-1:5000, ELISA 1:4000-1:20000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P01023 | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction