missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ alpha 1B-Glycoprotein Antibody (1H1), Novus Biologicals™

Product Code. 18346177 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346177 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346177 Supplier Novus Biologicals™ Supplier No. H00000001M15

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha 1B-Glycoprotein Monoclonal antibody specifically detects alpha 1B-Glycoprotein in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha 1B-Glycoprotein
Applications ELISA
Classification Monoclonal
Clone 1H1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH35719
Gene Alias Alpha-1-B glycoprotein;alpha1B-glycoprotein;Alpha-1B-glycoprotein;alpha-1-B-glycoprotein;DKFZp686F0970;epididymis secretory sperm binding protein Li 163pA;HEL-S-163pA
Host Species Mouse
Immunogen A1BG (AAH35719, 1 a.a. ∽ 373 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAPVSWITPGLKTTAVCRGVLRGVTFLLRREGDHEFLEVPEAQEDVEATFPVHQPGNYSCSYRTDGEGALSEPSATVTIEELAAPPPPVLMHHGESSQVLHPGNKVTLTCVAPLSGVDFQLRRGEKELLVPRSSTSPDRIFFHLNAVALGDGGHYTCRYRLHDNQNGWSGDSAPVELILSDETLPAPEFSPEPESGRALRLRCLAPLEGARFALVREDRGGRRVHRFQSPAGTEALFELHNISVADSANYSCVYVDLKPPFGGSAPSERLELHVDGPPPRPQLRATWSGAVLAGRDAVLRCEGPIPDVTFELLREGETKAVKTVRTPGAAANLELIFVGPQHAGNYRCRYRSWVPHTFESELSDPVELLVAES
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.