missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1A Adrenergic R/ADRA1A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£343.00 - £557.00
Specifications
| Antigen | alpha-1A Adrenergic R/ADRA1A |
|---|---|
| Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364533
|
Novus Biologicals
NBP3-17211-25UL |
25 μg |
£343.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329286
|
Novus Biologicals
NBP3-17211-100UL |
100 μg |
£557.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-1A Adrenergic R/ADRA1A Polyclonal antibody specifically detects alpha-1A Adrenergic R/ADRA1A in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| alpha-1A Adrenergic R/ADRA1A | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, GPCR, Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 148 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VFLSGNASDSSNCTQPPAPVNISKAI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title