missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1A Adrenergic R/ADRA1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16661-100UL
This item is not returnable.
View return policy
Description
alpha-1A Adrenergic R/ADRA1A Polyclonal antibody specifically detects alpha-1A Adrenergic R/ADRA1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| alpha-1A Adrenergic R/ADRA1A | |
| Polyclonal | |
| Western Blot, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human alpha-1A Adrenergic R/ADRA1A (P35348). | |
| 100 μg | |
| Apoptosis, GPCR, Neuroscience | |
| 148 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction