missing translation for 'onlineSavingsMsg'
Learn More

alpha 1-Microglobulin Antibody (3F1), Novus Biologicals™

Product Code. 18389777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389777 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389777 Supplier Novus Biologicals Supplier No. H00000259M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha 1-Microglobulin Monoclonal antibody specifically detects alpha 1-Microglobulin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha 1-Microglobulin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3F1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH41593
Gene Alias A1M, alpha-1-microglobulin/bikunin precursor, bikunin, EDC1, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI
Host Species Mouse
Immunogen AMBP (AAH41593, 19 a.a. ~ 352 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AGPVPTPPDNIQVQGNFNISRIYGKWYNLAIGSTCPWLKKIMDRMTVSTLVLGEGATEAEISMTSTRWRKGVCEETSGAYEKTDTDGKFLYHKSKWNITMESYVVHTNYDEYAIFLTKKFSRHHGPTITAKLYGRAPQLRETLLQDFRVVAQGVGIPEDSIFTMADRGECVPGEQEPEPILIPRVRRAVLPQEEEGSGGGQLVTEVTKKEDSCQLGYSAGPCMGMTSRYFYNGTSMACETFQYGGCMGNGNNFVTEKECLQTCRTVAACNLPIVRGPCRAFIQLWAFDAVKGKCVLFPYGGCQGNGNKFYSEKECREYCGVPGDGDEELLRFSN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.