missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1 Mannosidase 1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-977
This item is not returnable.
View return policy
Description
alpha 1 Mannosidase 1A Polyclonal specifically detects alpha 1 Mannosidase 1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| alpha 1 Mannosidase 1A | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| Alpha-1,2-mannosidase IA, EC 3.2.1, EC 3.2.1.113, HUMM3, HUMM9, Man(9)-alpha-mannosidase, MAN9, Man9-mannosidase, Mannosidase alpha class 1A member 1, mannosidase, alpha, class 1A, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IA, Processing alpha-1,2-mannosidase IA | |
| Rabbit | |
| 73 kDa | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 4121 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 0.7 mg/mL | |
| Western Blot 0.5-1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-10 ug/ml | |
| P33908 | |
| MAN1A1 | |
| Synthetic peptide corresponding to amino acides 138-154 of human alpha 1,2 Mannosidase IA, with N-terminal added cysteine, conjugated to KLH. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects alpha 1,2-Mannosidase IA. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction