missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha-1 Antitrypsin Monoclonal Antibody (1AT11-210.1), Invitrogen™

Product Code. 17810486
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Packungsgröße:
100µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
17810486 100 μg 100µg
11568912 500 μg 1mg
17830296 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17810486 Lieferant Invitrogen™ Lieferanten-Nr. MA546034

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

MA1-10626 detects alpha-1 Antitrypsin from human samples. MA1-10626 does not cross react with alpha-1-antichymotrysin. MA1-10626 has been successfully used in ELISA applications. The MA1-10626 immunogen is purified human serum alpha-1 Antitrypsin.

Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen alpha-1 Antitrypsin
Applications ELISA
Classification Monoclonal
Clone 1AT11-210.1
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SERPINA1
Gene Accession No. P01009
Gene Alias a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha 1 antitrypsin; alpha-1 antitrypsin; alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; protease inhibitor 1 (anti-elastase, antitrypsin, alpha-1); Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Protease 1; serine protease inhibitor 1-1; Serine protease inhibitor A1a; serine protease inhibitor alpha 1; serine proteinase inhibitor, clade A, member 1; serpin A1; Serpin A1a; serpin family A member 1; serpin peptidase inhibitor clade A member 1; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; SERPINA1; Serpina1a; Short peptide from AAT; SPAAT; Spi1; Spi1-1; Spi1-3; TRP1; TRY1
Gene Symbols SERPINA1
Host Species Mouse
Immunogen Purified human serum alpha-1 Antitrypsin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.