missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1 Antichymotrypsin Mouse anti-Human, Clone: 3F2F10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | alpha 1 Antichymotrypsin |
|---|---|
| Clone | 3F2F10 |
| Concentration | 0.77 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16877625
|
Proteintech
66078-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16867625
|
Proteintech
66078-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this proteiNULLs sequence have been implicated in Alzheimer′s disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease.Specifications
| alpha 1 Antichymotrypsin | |
| 0.77 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1 Antichymotrypsin, AACT, ACT, Alpha 1 antichymotrypsin, GIG24, GIG25, Serpin A3, SERPINA3 | |
| SERPINA3 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3F2F10 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01011 | |
| 12 | |
| alpha 1 Antichymotrypsin Fusion Protein Ag2830 | |
| Primary | |
| -20°C | |
| SERPINA3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title