missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1-Acid Glycoprotein Antibody (C10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43329
This item is not returnable.
View return policy
Description
alpha 1-Acid Glycoprotein Monoclonal antibody specifically detects alpha 1-Acid Glycoprotein in Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| alpha 1-Acid Glycoprotein | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| AGP 1, AGP1alpha-1-acid glycoprotein 1, AGP-A, OMD 1, ORM, orosomucoid 1, orosomucoid-1 | |
| Immunized with recombinant Rat alpha 1-Acid Glycoprotein (Uniprot#: P02764; Gln19∽Gln186) | |
| 100 μL | |
| Cancer, Immunology, Innate Immunity | |
| 5004 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| C10 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction