missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ alpha 1-Acid Glycoprotein Antibody (2F9-1F10), Novus Biologicals™

Product Code. 18345918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345918 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345918 Supplier Novus Biologicals™ Supplier No. H00005004M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha 1-Acid Glycoprotein Monoclonal antibody specifically detects alpha 1-Acid Glycoprotein in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha 1-Acid Glycoprotein
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 2F9-1F10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH26238
Gene Alias AGP 1, AGP1alpha-1-acid glycoprotein 1, AGP-A, OMD 1, ORM, orosomucoid 1, orosomucoid-1
Host Species Mouse
Immunogen ORM1 (AAH26238, 18 a.a. ∽ 201 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AQIPLCANLVPVPITNATLDRITGKWFYIASAFRNEEYNKSVQEIQATFFYFTPNKTEDTIFLREYQTRQDQCIYNTTYLNVQRENGTISRYVGGQEHFAHLLILRDTKTYMLAFDVNDEKNWGLSVYADKPETTKEQLGEFYEALDCLRIPKSDVVYTDWKKDKCEPLEKQHEKERKQEEGES
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5004
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.