missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpelisib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Alpelisib (BYL-719) is a potent, selective, and orally active PI3Kα inhibitor. Alpelisib (BYL-719) shows efficacy in targeting PIK3CA-mutated cancer. Alpelisib (BYL-719) also inhibits p110α/p110γ/p110δ/p110β with IC50s of 5/250/290/1200 nM, respectively. Antineoplastic activity.
Specifications
Specifications
| Chemical Name or Material | Alpelisib |
| CAS | 1217486-61-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H22F3N5O2S |
| Quantity | 10 mg |
| Synonym | BYL-719 |
| Solubility Information | DMSO : 83.33 mg/mL (188.76 mM; Need ultrasonic) |
| SMILES | CC(N=C(S1)NC(N2CCC[C@H]2C(N)=O)=O)=C1C3=CC(C(C)(C(F)(F)F)C)=NC=C3 |
| Molecular Weight (g/mol) | 441.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction