missing translation for 'onlineSavingsMsg'
Learn More

Alpaca monoclonal [2F3] Anti-human IgG1/IgG4 antibody (Alexa Fluor™ 647), Abcam

Product Code. 32112776 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112776 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112776 Supplier Abcam Supplier No. ab325308100uL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpaca Monoclonal Antibody

Recombinant secondary antibody suitable for ICC/IF. Ideal for fluorescent cell and tissue imaging.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen IGG1_HUMAN (IGG1)
Applications Immunocytochemistry/Immunofluorescence
Cellular Localization Secreted; Cell membrane
Classification Monoclonal
Clone '2F3
Conjugate Alexa Fluor 647
Formulation Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene IGG1_HUMAN
Gene Accession No. P0DOX5
Host Species Alpaca
Immunogen IGG1_HUMAN (IGG1)
Purification Method Size-exclusion chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -80°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene IGG1_HUMAN
Target Molecule IGG1_HUMAN (IGG1)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.