Learn More
Alpaca anti-Rabbit IgG Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand:  Invitrogen SA510326
Product Code. 17192224
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Biclonal nanobodies specific for rabbit IgG Fab and Fc. CTK0101: KD = 0.2 nM, CTK0102: KD = 1.2 nM Non Cross-Reactivity includes: Mouse, rat, sheep, goat, and guinea pig serum. Cross-reactivity to: Human and macaque serum Rapid workflow: Simultaneous incubation of rabbit IgG primary antibody and Alpaca anti-rabbit IgG Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Rabbit IgG Nano (VHH) Recombinant Monoclonal antibody specifically detects Rabbit IgG Nano (VHH) in Rabbit samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit IgG Nano (VHH)
Monoclonal
0.5 mg/mL
10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7
Alpaca
Affinity chromatography
RUO
Rabbit
Liquid
Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
CTK0101/CTK0102
Alexa Fluor 647
Biclonal; Nanobodies; recombinant VHH
Purified rabbit IgG
10 μL
Secondary
-20° C, Avoid Freeze/Thaw Cycles