Learn More
Alpaca anti-Rabbit IgG Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 488, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand: Invitrogen SA510322
Description
Biclonal nanobodies specific for rabbit IgG Fab and Fc. CTK0101: KD = 0.2 nM, CTK0102: KD = 1.2 nM Non Cross-Reactivity includes: Mouse, rat, sheep, goat, and guinea pig serum. Cross-reactivity to: Human and macaque serum Rapid workflow: Simultaneous incubation of rabbit IgG primary antibody and Alpaca anti-rabbit IgG Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Rabbit IgG Nano (VHH) Recombinant Monoclonal antibody specifically detects Rabbit IgG Nano (VHH) in Rabbit samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Specifications
| Rabbit IgG Nano (VHH) | |
| Monoclonal | |
| 0.5 mg/mL | |
| 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 | |
| Alpaca | |
| Affinity chromatography | |
| RUO | |
| Rabbit | |
| Liquid |
| Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot | |
| CTK0101/CTK0102 | |
| Alexa Fluor 488 | |
| Biclonal; Nanobodies; recombinant VHH | |
| Purified rabbit IgG | |
| 10 μL | |
| Secondary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.