Learn More
Alpaca anti-Mouse IgG2b Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand: Invitrogen SA510339
Description
Biclonal nanobodies specific for mouse IgG2b Fc. CTK0105: KD = 5 nM, CTK0106: KD = 0.2 nM Non Cross-Reactivity includes: Rabbit, rat, goat, sheep, guinea pig, human, and macaque serum and to mouse IgG1, IgG2a, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG2b primary antibody and Alpaca anti-mouse IgG2b Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek
Mouse IgG2b Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG2b Nano (VHH) in Mouse samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Specifications
| Mouse IgG2b Nano (VHH) | |
| Monoclonal | |
| 0.5 mg/mL | |
| 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 | |
| Alpaca | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Liquid |
| Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot | |
| CTK0105/CTK0106 | |
| Alexa Fluor 647 | |
| Biclonal; Fc-specific recombinant VHH; Nanobodies | |
| Purified mouse IgG2b | |
| 100 μL | |
| Secondary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.