Learn More
Alpaca anti-Mouse IgG2b Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 568, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand:  Invitrogen SA510336
Product Code. 17192234
  • £102.00 / 10µL
Description
Biclonal nanobodies specific for mouse IgG2b Fc. CTK0105: KD = 5 nM, CTK0106: KD = 0.2 nM Non Cross-Reactivity includes: Rabbit, rat, goat, sheep, guinea pig, human, and macaque serum and to mouse IgG1, IgG2a, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG2b primary antibody and Alpaca anti-mouse IgG2b Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek
Mouse IgG2b Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG2b Nano (VHH) in Mouse samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG2b Nano (VHH)
Monoclonal
0.5 mg/mL
10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7
Alpaca
Affinity chromatography
RUO
Mouse
Liquid
Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
CTK0105/CTK0106
Alexa Fluor 568
Biclonal; Fc-specific recombinant VHH; Nanobodies
Purified mouse IgG2b
10 μL
Secondary
-20° C, Avoid Freeze/Thaw Cycles
IgG2b