Learn More
Alpaca anti-Mouse IgG1 Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand: Invitrogen SA510332
Description
Biclonal nanobodies specific for mouse IgG1 Fc. CTK0103: KD =0.13 nM, CTK0104: KD = 0.63nM Non Cross-Reactivity includes: Goat, guinea pig, rabbit, rat, sheep, human, and macaque serum and to mouse IgG2a, IgG2b, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG1 primary antibody and Alpaca anti-mouse IgG1 Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Mouse IgG1 Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG1 Nano (VHH) in Mouse samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Specifications
| Mouse IgG1 Nano (VHH) | |
| Monoclonal | |
| 0.5 mg/mL | |
| 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 | |
| Alpaca | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Liquid |
| Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot | |
| CTK0103/CTK0104 | |
| Alexa Fluor 647 | |
| Biclonal; Fc-specific recombinant VHH; Nanobodies | |
| Purified mouse IgG1 | |
| 10 μL | |
| Secondary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.