Learn More
Alpaca anti-Mouse IgG1 Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 568, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Brand:  Invitrogen SA510331
Product Code. 17142234
  • £335.00 / 100µL
Description
Biclonal nanobodies specific for mouse IgG1 Fc. CTK0103: KD =0.13 nM, CTK0104: KD = 0.63nM Non Cross-Reactivity includes: Goat, guinea pig, rabbit, rat, sheep, human, and macaque serum and to mouse IgG2a, IgG2b, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG1 primary antibody and Alpaca anti-mouse IgG1 Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Mouse IgG1 Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG1 Nano (VHH) in Mouse samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG1 Nano (VHH)
Monoclonal
0.5 mg/mL
10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7
Alpaca
Affinity chromatography
RUO
Mouse
Liquid
Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
CTK0103/CTK0104
Alexa Fluor 568
Biclonal; Fc-specific recombinant VHH; Nanobodies
Purified mouse IgG1
100 μL
Secondary
-20° C, Avoid Freeze/Thaw Cycles
IgG1