missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALOX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146002
This item is not returnable.
View return policy
Description
Suggested positive control: human Peripheral Blood Leukocyte protein.
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Specifications
| ALOX5 | |
| Polyclonal | |
| Unconjugated | |
| ALOX5 | |
| 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; F730011J02; leukotriene A4 synthase; LOG5; LOW QUALITY PROTEIN: arachidonate 5-lipoxygenase; lox5; LOX-5; LOX5A; polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11689, 240 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P09917, P48999 | |
| ALOX5 | |
| A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction