missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALOX5 Monoclonal Antibody (D2), Invitrogen™
Mouse Monoclonal Antibody
£161.00 - £350.00
Specifications
| Antigen | ALOX5 |
|---|---|
| Clone | D2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17919162
|
Invitrogen™
MA550877 |
200 μL |
£350.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17960663
|
Invitrogen™
MA550878 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.Specifications
| ALOX5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; F730011J02; leukotriene A4 synthase; LOG5; LOW QUALITY PROTEIN: arachidonate 5-lipoxygenase; lox5; LOX-5; LOX5A; polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3 | |
| ALOX5 | |
| IgG2b κ | |
| Protein A/G | |
| ALOX5 |
| D2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P09917, P12527 | |
| 240, 25290 | |
| Recombinant Human 5-LO protein, Glu461-Ser671 (Accession #P09917). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title