missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALOX15 Monoclonal Antibody (OTI7H6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525891
This item is not returnable.
View return policy
Description
ALOX15 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
Specifications
| ALOX15 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16050, P39654, Q02759 | |
| Alox15 | |
| Full length human recombinant protein of ALOX15 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI7H6 | |
| Unconjugated | |
| Alox15 | |
| 12/15-lipoxygenase; 12/15-LO; 12/15-LOX; 12-LO; 12-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX-1; 15LOX-1; 15-LOX2; Alox12; Alox12l; Alox15; Arachidonate 12-lipoxygenase, leukocyte-type; arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; L-12LO; LOG15 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11687, 246, 81639 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction