missing translation for 'onlineSavingsMsg'
Learn More

ALOX12B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA554949

Additional Details : Weight : 0.02000kg

Product Code. 15712881

  • £431.00 / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: LYKLLIPHTR YTVQINSIGR AVLLNEGGLS AKGMSLGVEG FAGVMVRALS ELTYDSLYLP NDFVERGVQD LPGYYYRDDS LAVWNALEKY VTEI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

ALOX12B Polyclonal antibody specifically detects ALOX12B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

ALOX12B
Polyclonal
Unconjugated
ALOX12B
12R-lipoxygenase; 12R-LOX; Alox12b; alox12b {ECO:0000312; Aloxe2; arachidonate 12-lipoxygenase, 12R type; arachidonate 12-lipoxygenase, 12R-type; ARCI2; e-LOX 2; e-LOX2; epidermis-type lipoxygenase 12; Epidermis-type lipoxygenase 2; RGD:1305330}
Rabbit
Antigen affinity chromatography
RUO
242
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75342
ALOX12B
Recombinant protein corresponding to Human ALOX12B
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only