missing translation for 'onlineSavingsMsg'
Learn More

ALOX12B Antibody (3G12), Novus Biologicals™

Product Code. 18342558 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342558 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342558 Supplier Novus Biologicals Supplier No. H00000242M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALOX12B Monoclonal antibody specifically detects ALOX12B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX12B
Applications ELISA
Classification Monoclonal
Clone 3G12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001130
Gene Alias 12R-LOXEC 1.13.11.-, arachidonate 12-lipoxygenase, 12R type, arachidonate 12-lipoxygenase, 12R-type, EC 1.13.11.33,12R-lipoxygenase, Epidermis-type lipoxygenase 12
Host Species Mouse
Immunogen ALOX12B (NP_001130, 171 a.a. ~ 261 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NPNRPEWNGYIPGFPILINFKATKFLNLNLRYSFLKTASFFVRLGPMALAFKVRGLLDCKHSWKRLKDIRKIFPGKKSVVSEYVAEHWAED
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 242
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.