missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alosetron Hydrochloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Alosetron Hydrochloride |
| CAS | 122852-69-1 |
| Molecular Formula | C17 H18 N4 O . Cl H |
| Quantity | 50 mg |
| Synonym | 2,3,4,5-Tetrahydro-5-methyl-2-[(4-methyl-1H-imidazol-5-yl)methyl]-1H-pyrido[4,3-b]indol-1-one Hydrochloride (1:1), 1H-Pyrido[4,3-b]indol-1-one, 2,3,4,5-tetrahydro-5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-, monohydrochloride (9CI), Alosetron hydrochloride, GR 68755C |
| InChI Formula | InChI=1S/C17H18N4O.ClH/c1-11-13(19-10-18-11)9-21-8-7-15-16(17(21)22)12-5-3-4-6-14(12)20(15)2;/h3-6,10H,7-9H2,1-2H3,(H,18,19);1H |
| SMILES | Cl.Cc1[nH]cnc1CN2CCc3c(C2=O)c4ccccc4n3C |
| IUPAC Name | 5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-3,4-dihydropyrido[4,3-b]indol-1-one;hydrochloride |
| Molecular Weight (g/mol) | 330.81 |
| Formula Weight | 330.1247 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction