missing translation for 'onlineSavingsMsg'
Learn More

Alosetron Hydrochloride, MedChemExpress

Product Code. 16498178
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498178

Brand: Medchem Express HY70050C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alosetron Hydrochloride (GR 68755C) is a potent and highly selective serotonin 5-HT3 receptor antagonist.

Alosetron Hydrochloride is used for the research of irritable bowel syndrome (IBS). Alosetron blocks the fast 5HT3-mediated depolarisation of guinea-pig myenteric and submucosal neurons, with IC50 at ∼55 nM. Alosetron Hydrochloride attenuates the visceral nociceptive effect of rectal distension in conscious or anaesthetised dogs. Anti-inflammatory effects.

Specifications

Chemical Name or Material Alosetron Hydrochloride
CAS 122852-69-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H19ClN4O
Quantity 10 mg
Synonym GR 68755CGR 68755 HydrochlorideGR 68755X Hydrochloride
Solubility Information H2O : 33.33 mg/mL (100.75 mM; Need ultrasonic)
SMILES O=C(C(C1=C2C=CC=C1)=C(N2C)CC3)N3CC4=C(NC=N4)C.[H]Cl
Molecular Weight (g/mol) 330.81
Formula Weight 330.81
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.