missing translation for 'onlineSavingsMsg'
Learn More

Aloin, MedChemExpress

Product Code. 16441071
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16441071

Brand: Medchem Express HYN0123

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aloin(Aloin-A; Barbaloin-A) is a natural antitumor anthraquinone glycoside with iron chelating and non-atherogenic activities.

Aloin(Aloin-A

Specifications

Chemical Name or Material Aloin
CAS 1415-73-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22O9
Quantity 100 mg
Synonym Aloin-A Barbaloin-A
Solubility Information DMSO : 125 mg/mL (298.76 mM; Need ultrasonic)
SMILES O=C1C2=C(C=CC=C2O)[C@H]([C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C4=CC(CO)=CC(O)=C14
Molecular Weight (g/mol) 418.39
Formula Weight 418.39
Percent Purity 98.21%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.