missing translation for 'onlineSavingsMsg'
Learn More

Alogliptin Benzoate, MedChemExpress

Product Code. 16437996
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437996

Brand: Medchem Express HYA0023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes.

Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes.

Specifications

Chemical Name or Material Alogliptin Benzoate
CAS 850649-62-6
Color White
Purity Grade Notes Research
Molecular Formula C25H27N5O4
Quantity 100 mg
Synonym SYR 322
Solubility Information DMSO : 50 mg/mL (108.34 mM; Need ultrasonic)
SMILES O=C(C=C(N1C[C@H](N)CCC1)N(C2=O)CC3=C(C#N)C=CC=C3)N2C.O=C(O)C4=CC=CC=C4
Molecular Weight (g/mol) 461.51
Formula Weight 461.51
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.