missing translation for 'onlineSavingsMsg'
Learn More

Alofanib, MedChemExpress

Product Code. 16499887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499887 5 mg 5mg
16497386 100 mg 100mg
16483576 25 mg 25mg
16473437 50 mg 50mg
16407418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499887 Supplier Medchem Express Supplier No. HY17601
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Alofanib (RPT835) is a potent and selective allosteric inhibitor of fibroblast growth factor receptor 2 (FGFR2). Anticancer and antiangiogenic activity.

Alofanib (RPT835) is a potent and selective allosteric inhibitor of fibroblast growth factor receptor 2 (FGFR2). Anticancer and antiangiogenic activity.

Specifications

Chemical Name or Material Alofanib
CAS 1612888-66-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H15N3O6S
Quantity 5 mg
Synonym RPT835
Solubility Information DMSO : ≥ 30.1 mg/mL (72.81 mM)
SMILES O=C(O)C1=CC=CC(S(=O)(NC2=CC(C3=CC=CN=C3)=C(C)C=C2[N+]([O-])=O)=O)=C1
Molecular Weight (g/mol) 413.4
Formula Weight 413.4
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.