missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103596
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ALMS1.
THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.
Specifications
| ALMS1 | |
| Polyclonal | |
| Unconjugated | |
| ALMS1 | |
| Alms1; ALMS1, centrosome and basal body associated protein; ALSS; Alstrom syndrome 1; Alstrom syndrome 1 homolog; Alstrom syndrome protein 1; DKFZp686A118; DKFZp686D1828; KIAA0328 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 297408, 7840 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TCU4 | |
| ALMS1 | |
| A synthesized peptide derived from human ALMS1(Accession Q8TCU4), corresponding to amino acid residues L3548-L3598. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction