missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium chloride, 97%, Thermo Scientific™

Catalog Number 10232662
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18480-23-4
C21H20ClP
338.81
MFCD00031542
FKMJROWWQOJRJX-UHFFFAOYSA-M
allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
2734120
triphenyl(prop-2-en-1-yl)phosphanium chloride
[Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
18480-23-4
C21H20ClP
338.81
MFCD00031542
FKMJROWWQOJRJX-UHFFFAOYSA-M
allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
2734120
triphenyl(prop-2-en-1-yl)phosphanium chloride
[Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 226°C to 230°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2CCHCH2P(C6H5)3Cl
Quantity 5 g
Formula Weight 338.82
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Allyltriphenylphosphonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Allyl triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'