missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethylsilane, 98+%

Product Code. 11364609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364609 250 g 250g
11489387 10 g 10g
11497626 10 g 10g
11407636 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11364609 Supplier Thermo Scientific Alfa Aesar Supplier No. A14662.30
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 762-72-1 | C6H14Si | 114.263 g/mol

Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-72-1
Molecular Formula C6H14Si
Molecular Weight (g/mol) 114.263
MDL Number MFCD00008635
InChI Key HYWCXWRMUZYRPH-UHFFFAOYSA-N
Synonym allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
PubChem CID 69808
IUPAC Name trimethyl(prop-2-enyl)silane
SMILES C[Si](C)(C)CC=C

Specifications

Boiling Point 85°C to 86°C
Quantity 250 g
UN Number UN1993
Beilstein 906755
Flash Point −10°C (14°F)
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 114.27
Percent Purity ≥98%
Refractive Index 1.408
Density 0.717
Chemical Name or Material Allyltrimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.