missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethoxysilane, 97+%

Product Code. 10734164 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734164 5 mL Glass bottle 5mL
10288843 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734164 Supplier Thermo Scientific Acros Supplier No. 388940050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2551-83-9 | C6H14O3Si | 162.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2551-83-9
Molecular Formula C6H14O3Si
Molecular Weight (g/mol) 162.26
MDL Number MFCD00053865
InChI Key LFRDHGNFBLIJIY-UHFFFAOYSA-N
Synonym allyltrimethoxysilane, silane, trimethoxy-2-propenyl, silane, trimethoxy-2-propen-1-yl, allyl trimethoxy silane, silane, allyltrimethoxy, allytrimethoxysilane, acmc-209gkn, trimethoxy prop-2-enyl silane, silane,trimethoxy-2-propen-1-yl
PubChem CID 75698
IUPAC Name trimethoxy(prop-2-enyl)silane
SMILES CO[Si](CC=C)(OC)OC

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 146°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2Si(OCH3)3
Refractive Index 1.4040 to 1.406
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: reacts
Formula Weight 162.26
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Allyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.