missing translation for 'onlineSavingsMsg'
Learn More

Allyltriethoxysilane, 97%

Product Code. 11336886
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336886 25 g 25g
11326886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336886 Supplier Thermo Scientific Alfa Aesar Supplier No. L04297.14
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2550-04-1 | C9H20O3Si | 204.34 g/mol

Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-04-1
MDL Number MFCD00009064

Specifications

Boiling Point 176°C to 177°C
Quantity 25 g
Molecular Formula C3H6Cl4Si
Linear Formula CH2=CHCH2Si(OCH2CH3)3
UN Number UN1993
Beilstein 1772306
Flash Point 21°C (69°F)
Synonym 3-chloropropyltrichlorosilane, trichloro 3-chloropropyl silane, 3-chloropropyl trichlorosilane, chloropropyltrichlorosilane, silane, trichloro 3-chloropropyl, silane, 3-chloropropyltrichloro, trichloro-3-chloro-propyl-silane, silane, trichloro chloropropyl, g-chloropropyltrichlorosilane, gamma-chloropropyl trichlorosilane
Solubility Information Hydrolyzes in water.
InChI Key OOXSLJBUMMHDKW-UHFFFAOYSA-N
SMILES C(C[Si](Cl)(Cl)Cl)CCl
IUPAC Name trichloro(3-chloropropyl)silane
Molecular Weight (g/mol) 211.97
PubChem CID 75693
Formula Weight 204.34
Percent Purity 97%
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.953
Chemical Name or Material Allyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.