missing translation for 'onlineSavingsMsg'
Learn More

Allyltrichlorosilane, 95%

Product Code. 11322276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322276 5 g 5g
11332276 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322276 Supplier Thermo Scientific Alfa Aesar Supplier No. L03703.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-37-9 | C3H5Cl3Si | 175.508 g/mol

Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

Solubility
Soluble in THF, toluene, benzene. Reacts with water.

Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-37-9
Molecular Formula C3H5Cl3Si
Molecular Weight (g/mol) 175.508
MDL Number MFCD00000486
InChI Key HKFSBKQQYCMCKO-UHFFFAOYSA-N
Synonym allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
PubChem CID 7867
IUPAC Name trichloro(prop-2-enyl)silane
SMILES C=CC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 117°C to 118°C
Quantity 5 g
Linear Formula CH2=CHCH2SiCl3
UN Number UN1724
Beilstein 1743449
Flash Point 18°C (64°F)
Solubility Information Soluble in THF,toluene,benzene. Reacts with water.
Formula Weight 175.52
Percent Purity 95%
Refractive Index 1.449
Sensitivity Moisture sensitive
Density 1.211
Chemical Name or Material Allyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.