missing translation for 'onlineSavingsMsg'
Learn More

Allylpalladium(II) chloride dimer, Pd 56.0% min

Product Code. 11366707
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366707 5 g 5g
11356707 0.25 g 0.25g
11346707 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366707 Supplier Thermo Scientific Alfa Aesar Supplier No. 010005.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12012-95-2 | C6H10Cl2Pd2 | 365.89 g/mol

Allylpalladium(II) chloride dimer is used as an important catalyst in Heck reaction. It reacts with cyclopentadienyl anion to give cyclopentadienyl allyl palladium. It acts as a precatalyst for asymmetric and cross-coupling catalysis. Further, it is used in the preparation of 1,4-diallyl-1,2-dihydroisoquinolines. It is also employed as a catalyst for greener Buchwald-Hartwig coupling reaction and involved in the synthesis of cationic palladium catalysts. In addition, it takes part in preparation of -heterocyclic carbene-palladium-eta3-allyl chloride complex, which is an efficient catalyst for the Suzuki-Miyaura cross-coupling reactions in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12012-95-2
Molecular Formula C6H10Cl2Pd2
Molecular Weight (g/mol) 365.89
MDL Number MFCD00044874
InChI Key PENAXHPKEVTBLF-UHFFFAOYSA-L
Synonym allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PubChem CID 61538
IUPAC Name chloropalladium(1+);prop-1-ene
SMILES Cl[Pd].Cl[Pd].c:c:c.c:c:c

Specifications

Melting Point 120°C (decomposition)
Color Yellow
Quantity 5 g
Beilstein 4124623
Sensitivity Air and moisture sensitive
Solubility Information Very soluble in dichloromethane and dichloroethane; Slightly soluble in toluene,insoluble in waterSoluble in chloroform,benzene,acetone and dichloromethane,dichloroethane and methanol; Slightly soluble in toluene; Insoluble in water.
Formula Weight 365.85
Concentration or Composition (by Analyte or Components) Pd 56.0% min.
Physical Form Powder
Chemical Name or Material Allylpalladium(II) chloride dimer

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.