Learn More
Allylpalladium(II) chloride dimer, Pd 56.0% min
Description
Allylpalladium(II) chloride dimer is used as an important catalyst in Heck reaction. It reacts with cyclopentadienyl anion to give cyclopentadienyl allyl palladium. It acts as a precatalyst for asymmetric and cross-coupling catalysis. Further, it is used in the preparation of 1,4-diallyl-1,2-dihydroisoquinolines. It is also employed as a catalyst for greener Buchwald-Hartwig coupling reaction and involved in the synthesis of cationic palladium catalysts. In addition, it takes part in preparation of -heterocyclic carbene-palladium-eta3-allyl chloride complex, which is an efficient catalyst for the Suzuki-Miyaura cross-coupling reactions in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 120°C (decomposition) |
| Color | Yellow |
| Quantity | 1 g |
| Beilstein | 4124623 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Very soluble in dichloromethane and dichloroethane; Slightly soluble in toluene,insoluble in waterSoluble in chloroform,benzene,acetone and dichloromethane,dichloroethane and methanol; Slightly soluble in toluene; Insoluble in water. |
| Formula Weight | 365.85 |
| Concentration or Composition (by Analyte or Components) | Pd 56.0% min. |
| Physical Form | Powder |
| Chemical Name or Material | Allylpalladium(II) chloride dimer |
Safety and Handling
- Allylpalladium(II) chloride dimer
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Allyl palladium chloride dimer
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.