missing translation for 'onlineSavingsMsg'
Learn More

Allyloxytrimethylsilane, 98%

Product Code. 15490258
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490258 100 g 100g
15470258 5 g 5g
15480258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490258 Supplier Thermo Scientific Alfa Aesar Supplier No. H53443.22
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18146-00-4 | C6H14OSi | 130.262 g/mol

Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18146-00-4
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00008643
InChI Key MNMVKGDEKPPREK-UHFFFAOYSA-N
Synonym allyloxytrimethylsilane, allyl trimethylsilyl ether, silane, trimethyl 2-propenyloxy, trimethyl 2-propenyloxy silane, o-trimethylsilylallyl alcohol, trimethylallyloxysilane, allyloxy trimethylsilane, silane, trimethyl 2-propen-1-yloxy, trimethyl prop-2-en-1-yloxy silane, acmc-1boq9
PubChem CID 87471
IUPAC Name trimethyl(prop-2-enoxy)silane
SMILES C[Si](C)(C)OCC=C

Specifications

Boiling Point 101°C to 102°C
Quantity 100 g
Linear Formula CH2=CHCH2OSi(CH3)3
UN Number UN1993
Beilstein 1739517
Flash Point 1°C (34°F)
Formula Weight 130.26
Percent Purity 99%
Chemical Name or Material Allyloxytrimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.