missing translation for 'onlineSavingsMsg'
Learn More

Allyloxy-tert-butyldimethylsilane, 98%, Thermo Scientific Chemicals

£26.00 - £277.00

Chemical Identifiers

CAS 105875-75-0
Molecular Formula C9H20OSi
MDL Number MFCD00239355
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15401218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55077.AA
5 mL
£26.00
5mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15411218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55077.AC
25 mL
£90.20
25mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15421218
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55077.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.50 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

105875-75-0
C9H20OSi
MFCD00239355

Specifications

120°C (50mmHg)
C9H20OSi
MFCD00239355
33°C (91°F)
172.34
1.421
Allyloxy-tert-butyldimethylsilane
105875-75-0
H2C=CHCH2OSi(CH3)2C(CH3)3
UN1993
Sparingly soluble in water (0.50g/L at 25°C).
98%
0.811
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only