missing translation for 'onlineSavingsMsg'
Learn More

Allyloxy-tert-butyldimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 15401218
Click to view available options
Quantity:
5 mL
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.50 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105875-75-0
Molecular Formula C9H20OSi
MDL Number MFCD00239355

Specifications

Boiling Point 120°C (50mmHg)
Quantity 5 mL
Linear Formula H2C=CHCH2OSi(CH3)2C(CH3)3
UN Number UN1993
Flash Point 33°C (91°F)
Solubility Information Sparingly soluble in water (0.50g/L at 25°C).
Formula Weight 172.34
Percent Purity 98%
Refractive Index 1.421
Density 0.811
Chemical Name or Material Allyloxy-tert-butyldimethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'