missing translation for 'onlineSavingsMsg'
Learn More

Allylmagnesium chloride, 1M in MeTHF

Product Code. 15488057
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488057 500 mL 500mL
15478057 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488057 Supplier Thermo Scientific Alfa Aesar Supplier No. H51170.AP
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2622-05-1 | C3H5ClMg | 100.83 g/mol

Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5?-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5′-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-05-1
Molecular Formula C3H5ClMg
Molecular Weight (g/mol) 100.83
MDL Number MFCD00000473
InChI Key PLYLAFITHJUEGX-UHFFFAOYSA-M
Synonym allylmagnesium chloride, magnesium, chloro-2-propenyl, magnesium, chloro-2-propen-1-yl, chloro prop-2-en-1-yl magnesium, allylmagnesium chloride solution, 2.0 m in thf, sjpdadpprujp@, allylmagnesiumchloride, allyl magnesium chloride, allyl-magnesium chloride, 2-propenyl magnesium chloride
PubChem CID 75804
IUPAC Name magnesium;prop-1-ene;chloride
SMILES Cl[Mg]CC=C

Specifications

Linear Formula CH2=CHCH2MgCl
Quantity 500 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 100.83
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Allylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.